Learn More
Aviva Systems Biology Corp ATG10 Antibody - C-terminal region (ARP54274_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54274_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATG10. Alternate names: APG10, APG10L, DKFZP586I0418, FLJ13954, pp12616ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61154