Learn More
Aviva Systems Biology Corp Atg12 Antibody - C-terminal region (ARP59030_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59030_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atg12. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Apg12l, MGC125080ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-286-563