Learn More
Aviva Systems Biology Corp ATG12 Antibody - middle region (ARP54265_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54265_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG12. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATG12. Alternate names: APG12, APG12L, FBR93, HAPG12ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-899