Learn More
Aviva Systems Biology Corp ATG13 Antibody - N-terminal region (ARP64384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG13. It was validated on Western Blot. . Alternate names: ATG13, FLJ20698, KIAA0652ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-252