Learn More
Sino Biological Recombinant Human Nck1 Protein (Full Length, His Tag)
Catalog No. 502574071
Encompass_Preferred
Recombinant full length human NCK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-4071 Supplier Sino Biological Supplier No. N0630H100
Recombinant full length human NCK was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.