Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci ATG16 Peptide
SDP

Catalog No. 16919554
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-919-554 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-919-554 Supplier ProSci Supplier No. 4427P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

ATG16 peptide is used for blocking the activity of ATG16 antibody.

Specifications

Gene ID (Entrez) 55054
Species Human
Accession Number NP_110430
Common Name ATG16 autophagy related 16-like 1 (S. cerevisiae)
Concentration 200 μg/mL
Content And Storage Store ATG16 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol ATG16L1
Quantity 0.05 mg
Sequence 18 amino acids near the center of human ATG16.
Synonym IBD10, WDR30, APG16L, ATG16A, ATG16L, UNQ9393/PRO34307, Autophagy-related protein 16-1, APG16-like 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.