Learn More
Aviva Systems Biology Corp ATG16L1 Antibody - N-terminal region (ARP58924_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58924_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG16L1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ATG16L1. Alternate names: APG16L, ATG16L, FLJ00045, FLJ10035, FLJ10828, FLJ22677, IBD10, WDR30, ATG16AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-529