Learn More
Aviva Systems Biology Corp ATG4A Antibody - N-terminal region (ARP42689_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42689_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATG4A. Alternate names: APG4A, AUTL2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54029