Learn More
Aviva Systems Biology Corp ATG4B Antibody - C-terminal region (ARP34434_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34434_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG4B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATG4B. Alternate names: APG4B, AUTL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55394