Learn More
Aviva Systems Biology Corp ATG4B Antibody - N-terminal region (ARP58929_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58929_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG4B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ATG4B. Alternate names: APG4B, AUTL1, MGC1353ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-531