Learn More
Aviva Systems Biology Corp ATG4C Antibody - middle region (ARP58863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG4C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATG4C. Alternate names: APG4-C, APG4C, AUTL1, AUTL3, FLJ14867ApplicationWB. Species reactivity: Cow, Dog, Horse, Human
Catalog No. 50-286-508