Learn More
Aviva Systems Biology Corp ATG4D Antibody - middle region (ARP58865_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58865_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG4D. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATG4D. Alternate names: APG4-D, APG4D, AUTL4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit
Catalog No. 50-286-509