Learn More
Aviva Systems Biology Corp ATG5 Antibody - C-terminal region (ARP54267_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54267_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ATG5. Alternate names: APG5, APG5L, ASP, hAPG5, APG5-LIKEApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61150