Learn More
Aviva Systems Biology Corp ATG5 Antibody - middle region (ARP59071_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59071_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATG5. Alternate names: APG5, APG5-LIKE, APG5L, ASP, hAPG5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-571