Learn More
Cayman Chemical ATG5 MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502939832
Encompass_Preferred
Immunogen: Full-length human recombinant ATG5 protein • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (-) ATG8 • Applications: ELISA, IF
Catalog No. 50-293-9832 Supplier Cayman Chemical Supplier No. 28812100 UG
Immunogen: Full-length human recombinant ATG5 protein • Host: Mouse • Species Reactivity: (+) Human • Cross Reactivity: (-) ATG8 • Applications: ELISA, IF