Learn More
Cayman Chemical ATG5 MoncLNal ANTIBDYClo 100ug
SDP
Catalog No. 502934524
Encompass_Preferred
Immunogen: Full-length human recombinant ATG5 protein • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (-) ATG8 • Applications: ELISA, IHC, and WB • MW: ~56 kDa for the ATG5-ATG12 complex
Catalog No. 50-293-4524 Supplier Cayman Chemical Supplier No. 26671100 UG
Immunogen: Full-length human recombinant ATG5 protein • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (-) ATG8 • Applications: ELISA, IHC, and WB • MW: ~56 kDa for the ATG5-ATG12 complex