Learn More
Medchemexpress LLC Atg7-IN-2 | 2226227-75-2 | 99.3% | 376.35 | 50 MG
SDP
Catalog No. 5000359820
Encompass_Preferred
ATG7-IN-2 (compound 1) is a potent ATG7 inhibitor, with an IC50 of 0.089 μM. It inhibits the autophagy marker LC3B. This product is for research use only and is not sold to patients.
  • Potent ATG7 inhibitor with an IC50 of 0.089 μM.
  • Inhibits autophagy marker LC3B.
  • Inhibits the formation of the ATG7-ATG8 thioester in HEK293 cells with an IC50 of 0.335 μM.
  • Suppresses LC3B lipidation in H4 cells with an IC50 of 2.6 μM.
  • Reduces cell viability in H1650 cells with an EC50 of 2.6 μM.
Catalog No. 50-003-59820 Supplier Medchemexpress LLC Supplier No. HY14613050MG
ATG7-IN-2 (compound 1) is a potent ATG7 inhibitor, with an IC50 of 0.089 μM. It inhibits the autophagy marker LC3B. This product is for research use only and is not sold to patients.
  • Potent ATG7 inhibitor with an IC50 of 0.089 μM.
  • Inhibits autophagy marker LC3B.
  • Inhibits the formation of the ATG7-ATG8 thioester in HEK293 cells with an IC50 of 0.335 μM.
  • Suppresses LC3B lipidation in H4 cells with an IC50 of 2.6 μM.
  • Reduces cell viability in H1650 cells with an EC50 of 2.6 μM.