Learn More
Aviva Systems Biology Corp Atg9a Antibody - C-terminal region (ARP58930_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58930_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atg9a. It was validated on Western Blot. . Alternate names: AU019532, Apg9l1, Atg9, Atg9l1, MGC105176ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-532