Learn More
Aviva Systems Biology Corp ATG9B Antibody - C-terminal region (ARP58867_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58867_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATG9B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ATG9B. Alternate names: APG9L2, NOS3AS, SONEApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-510