Learn More
Apexbio Technology LLC Atglistatin,10mg CAS# 1469924-27-3
SDP
Supplier:  Apexbio Technology LLC B302110
Encompass_Preferred
Atglistatin is a potent and selective inhibitor of ATGL with IC50 value of 0.7 μM [1]. Adipose triglyceride lipase (ATGL) is a rate-limiting enzyme in the mobilization of fatty acids from cellular triglyceride stores. Deregulated fatty acid metabolism may lead to metabolic disorders and dyslipidemic [1]. Atglistatin is a potent and selective ATGL inhibitor. In lysates from E. coli overexpressing ATGL, atglistatin inhibited ATGL activity with IC50 value of 0.7 μM. Atglistatin at concentrations up to 50 μM had no cytotoxicity. Also, atglistatin inhibited ATGL with Ki value of 355 nM in a competitive way. In white adipose tissue (WAT) lysates of wild-type mice, atglistatin inhibited triacylglycerol (TG) hydrolase activity by 78%. The combination of Atglistatin and the HSL inhibitor Hi 76-007917 inhibited TG hydrolase activity by 95%. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0126