Learn More
Cayman Chemical Atglistatin, 1469924-27-3, 1 mg
SDP
Supplier:  Cayman Chemical 152841
Encompass_Preferred
A potent, selective, and competitive inhibitor of ATGL (IC50 = 0.7 μM); does not inhibit hormone-sensitive lipase, monoglyceride lipase, pancreatic lipase, lipoprotein lipase, or other lysophospholipases; blocks lipolysis by ATGL in vitro, in white adipose tissue organ cultures, and in vivo
Catalog No. NC0843366