Learn More
Medchemexpress LLC HY-15859 5mg Medchemexpress, Atglistatin CAS:1469924-27-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158595MG
Encompass_Preferred
Medchemexpress, HY-15859 5mg Atglistatin CAS:1469924-27-3 Atglistatin is a selective adipose triglyceride lipase (ATGL) inhibitor which inhibits lipolysis with an IC50 of 0.7 μM in vitro. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9176