Learn More
Medchemexpress LLC ATH686 | 853299-52-2 | 99.5% | 515.53 | 50 MG
SDP
Catalog No. 5000361859
Encompass_Preferred
ATH686 is a potent, selective, and ATP-competitive FLT3 inhibitor. It targets mutant FLT3 protein kinase activity, inhibiting the proliferation of cells harboring FLT3 mutants by inducing apoptosis and cell cycle inhibition. It has demonstrated antileukemic effects.
  • Potent, selective, and ATP-competitive FLT3 inhibitor.
  • Targets mutant FLT3 protein kinase activity.
  • Induces apoptosis and cell cycle inhibition.
  • Inhibits proliferation of cells harboring FLT3 mutants.
  • Demonstrates antileukemic effects.
  • For research use only.
Catalog No. 50-003-61859 Supplier Medchemexpress LLC Supplier No. HY1500350MG
ATH686 is a potent, selective, and ATP-competitive FLT3 inhibitor. It targets mutant FLT3 protein kinase activity, inhibiting the proliferation of cells harboring FLT3 mutants by inducing apoptosis and cell cycle inhibition. It has demonstrated antileukemic effects.
  • Potent, selective, and ATP-competitive FLT3 inhibitor.
  • Targets mutant FLT3 protein kinase activity.
  • Induces apoptosis and cell cycle inhibition.
  • Inhibits proliferation of cells harboring FLT3 mutants.
  • Demonstrates antileukemic effects.
  • For research use only.