Learn More
Medchemexpress LLC ATH686 | 853299-52-2 | 99.54% | 515.53 | 5 MG
SDP
Catalog No. 5000361273
Encompass_Preferred
ATH686 is a potent, selective, and ATP-competitive FLT3 inhibitor that targets mutant FLT3 protein kinase activity. It inhibits the proliferation of cells harboring FLT3 mutants by inducing apoptosis and cell cycle inhibition, also exhibiting antileukemic effects.
  • Potently inhibits cell proliferation (IC50 around 0.001 μM) in FLT3-ITD-Ba/F3 cells and D835Y-Ba/F3 cells
  • Induces apoptosis
  • Inhibits autophosphorylation of mutant FLT3
  • Exhibits antileukemic effects
Catalog No. 50-003-61273 Supplier Medchemexpress LLC Supplier No. HY150035MG
ATH686 is a potent, selective, and ATP-competitive FLT3 inhibitor that targets mutant FLT3 protein kinase activity. It inhibits the proliferation of cells harboring FLT3 mutants by inducing apoptosis and cell cycle inhibition, also exhibiting antileukemic effects.
  • Potently inhibits cell proliferation (IC50 around 0.001 μM) in FLT3-ITD-Ba/F3 cells and D835Y-Ba/F3 cells
  • Induces apoptosis
  • Inhibits autophosphorylation of mutant FLT3
  • Exhibits antileukemic effects