Learn More
Medchemexpress LLC Ath686 | 853299-52-2 | MFCD22665713 | 99.5% | 515.53 | C25H28F3N7O2 | 10 MG
SDP
Catalog No. 5000285856
Encompass_Preferred
ATH686 is a potent, selective, ATP-competitive FLT3 kinase inhibitor used in research to study FLT3-mutant leukemia. It inhibits mutant FLT3 activity, induces apoptosis, and suppresses proliferation in FLT3-mutant cells.
  • Potent, selective ATP-competitive FLT3 inhibition.
  • Demonstrated induction of apoptosis and cell-cycle arrest in FLT3-mutant cells.
  • High purity suitable for biochemical and cellular assays.
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-85856 Supplier Medchemexpress LLC Supplier No. HY1500310MG
ATH686 is a potent, selective, ATP-competitive FLT3 kinase inhibitor used in research to study FLT3-mutant leukemia. It inhibits mutant FLT3 activity, induces apoptosis, and suppresses proliferation in FLT3-mutant cells.
  • Potent, selective ATP-competitive FLT3 inhibition.
  • Demonstrated induction of apoptosis and cell-cycle arrest in FLT3-mutant cells.
  • High purity suitable for biochemical and cellular assays.
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Stable under recommended storage conditions for long-term use.