Learn More
Athens Research And Technology Inc Inter Alpha Inhibitor Proteins (IαIp), Native Human Plasma, ≥90% (SDS-PAGE), 100 µg
Catalog No. NC3995198
Encompass_Preferred
Inter Alpha Inhibitor Proteins (IαIp) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied frozen in a 100 µg pack. Molecular weight: 225 kDa and 125 kDa.
Catalog No. NC3995198 Supplier Athens Research And Technology Inc Supplier No. 1616090109100UG
Inter Alpha Inhibitor Proteins (IαIp) is a native protein purified from human plasma with ≥90% (SDS-PAGE), supplied frozen in a 100 µg pack. Molecular weight: 225 kDa and 125 kDa.