Learn More
AdipoGen Athidathion
SDP
Supplier:  AdipoGen CDXA0265M100
Encompass_Preferred
Chemical. CAS 19691-80-6. Formula C8H15N2O4PS3. MW 330.39. Synthetic. An organophosphate pesticide. Insecticide and acaricide. Acetylcholinesterase AChE inhibitor in the nervous system. Used in the past for the control of sucking and chewing insects. Compound can be used as analytical reference material.
Catalog No. 50-256-1774