Learn More
Med Vet International Athroscopic Stifle Lever 280mm Teflon Handle, J0444Q
Catalog No. 5000584443
Encompass_Preferred
Exposure of the medial meniscus for examination and treatment is never easy. Many instruments have been developed for the purpose. The original technique required a Hohman to advance the tibia. Standard Hohmans are not designed for the purpose and can cause trauma to both tibia and trochlea. The original stifle lever addresses these issues but is a little bulky for arthroscopic use. The arthroscopic version is finer, longer, and has a substantial Teflon handle allowing better manipulation of the joint.
Catalog No. 50-005-84443 Supplier Med Vet International Supplier No. JOR444Q
Exposure of the medial meniscus for examination and treatment is never easy. Many instruments have been developed for the purpose. The original technique required a Hohman to advance the tibia. Standard Hohmans are not designed for the purpose and can cause trauma to both tibia and trochlea. The original stifle lever addresses these issues but is a little bulky for arthroscopic use. The arthroscopic version is finer, longer, and has a substantial Teflon handle allowing better manipulation of the joint.