Learn More
Medchemexpress LLC HY-P0172A 1mg Medchemexpress, ATI-2341 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0172A1MG
Encompass_Preferred
Medchemexpress, HY-P0172A 1mg ATI-2341 (TFA) CAS: ATI-2341 is a potent and functionally selective allosteric agonist of C-X-C chemokine receptor type 4 (CXCR4), which functions as a biased ligand, favoring Gαi activation over Gα13. ATI-2341 activates the inhibitory heterotrimeric G protein (Gi) to promote inhibition of cAMP production and induce calcium mobilization. ATI-2341 is a potent and efficacious mobilizer of bone marrow polymorphonuclear neutrophils (PMNs) and hematopoietic stem and progenitor cells (HSPCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0517