Learn More
Medchemexpress LLC ATI-2341 TFA | 98.1% | 2256.82 (free base) | 5 MG
SDP
Catalog No. 5000426435
Encompass_Preferred
ATI-2341 is a potent and functionally selective allosteric agonist of C-X-C chemokine receptor type 4 (CXCR4). This compound activates the inhibitory heterotrimeric G protein (Gi) to promote inhibition of cAMP production and induce calcium mobilization.
  • Acts as a biased ligand, favoring Gαi activation over Gα13
  • Potent and efficacious mobilizer of bone marrow polymorphonuclear neutrophils (PMNs)
  • Potent and efficacious mobilizer of hematopoietic stem and progenitor cells (HSPCs)
Catalog No. 50-004-26435 Supplier Medchemexpress LLC Supplier No. HYP0172A5MG
ATI-2341 is a potent and functionally selective allosteric agonist of C-X-C chemokine receptor type 4 (CXCR4). This compound activates the inhibitory heterotrimeric G protein (Gi) to promote inhibition of cAMP production and induce calcium mobilization.
  • Acts as a biased ligand, favoring Gαi activation over Gα13
  • Potent and efficacious mobilizer of bone marrow polymorphonuclear neutrophils (PMNs)
  • Potent and efficacious mobilizer of hematopoietic stem and progenitor cells (HSPCs)