Learn More
Aviva Systems Biology Corp ATIC Antibody - middle region (ARP46094_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46094_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ATIC. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ATIC. Alternate names: AICAR, AICARFT, FLJ93545, IMPCHASE, PURHApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59222