Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atic-in-1 acetate | 1402453-15-9 | 100.0% | 523.58 | C23H37N7O7 | 10 MG
SDP

Catalog No. 5000286221
Encompass_Preferred

ATIC-IN-1 acetate is a small-molecule inhibitor that targets ATIC dimerization, interfering with de novo purine biosynthesis (Ki = 685 nM). It has demonstrated anti-tumor activity by reducing cell numbers and proliferation and is supplied for in vitro and in vivo research applications.

  • Inhibits ATIC dimerization (Ki = 685 nM).
  • Demonstrated anti-tumor activity in cell-based assays.
  • High purity: 99.96%.
  • Molecular formula C23H37N7O7; molecular weight 523.58.
  • Soluble in DMSO and water at 50 mg/mL with ultrasonic assistance.
  • Stable under sealed storage; 4°C recommended; in solvent, -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-86221 Supplier Medchemexpress LLC Supplier No. HY150252A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ATIC-IN-1 acetate is a small-molecule inhibitor that targets ATIC dimerization, interfering with de novo purine biosynthesis (Ki = 685 nM). It has demonstrated anti-tumor activity by reducing cell numbers and proliferation and is supplied for in vitro and in vivo research applications.

  • Inhibits ATIC dimerization (Ki = 685 nM).
  • Demonstrated anti-tumor activity in cell-based assays.
  • High purity: 99.96%.
  • Molecular formula C23H37N7O7; molecular weight 523.58.
  • Soluble in DMSO and water at 50 mg/mL with ultrasonic assistance.
  • Stable under sealed storage; 4°C recommended; in solvent, -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.