Learn More
Medchemexpress LLC ATIC-IN-1 acetate | 3067165-96-9 | C23H37N7O7 | 1 ML
SDP
Catalog No. 5000375580
Encompass_Preferred
ATIC-IN-1 acetate, also referred to as compound 14, is a specific inhibitor targeting the dimerization of Aminoimidazole carboxamide ribonucleotide transformylase/inosine monophosphate cyclohydrolase (ATIC). This dimerization is critical for the AICAR transformylase activity of ATIC, an enzyme involved in de novo purine biosynthesis. It demonstrates anti-tumor activity by reducing cell numbers and cell division rates.
  • Inhibits ATIC dimerization crucial for AICAR transformylase activity
  • Ki value of 685 nM
  • Exhibits anti-tumor activity by reducing cell numbers
  • Reduces cell division rates
  • Specific inhibitor at 10 μM and 50 μM
  • Inhibits proliferation of MCF-7 cells at 100-500 μM over 48 hours
Catalog No. 50-003-75580 Supplier Medchemexpress LLC Supplier No. HY150252A10MM/1ML
ATIC-IN-1 acetate, also referred to as compound 14, is a specific inhibitor targeting the dimerization of Aminoimidazole carboxamide ribonucleotide transformylase/inosine monophosphate cyclohydrolase (ATIC). This dimerization is critical for the AICAR transformylase activity of ATIC, an enzyme involved in de novo purine biosynthesis. It demonstrates anti-tumor activity by reducing cell numbers and cell division rates.
  • Inhibits ATIC dimerization crucial for AICAR transformylase activity
  • Ki value of 685 nM
  • Exhibits anti-tumor activity by reducing cell numbers
  • Reduces cell division rates
  • Specific inhibitor at 10 μM and 50 μM
  • Inhibits proliferation of MCF-7 cells at 100-500 μM over 48 hours