Learn More
Medchemexpress LLC ATIC-IN-1 acetate | 3067165-96-9 | C23H37N7O7 | 50 MG
SDP
Catalog No. 5000375569
Encompass_Preferred
ATIC-IN-1 acetate is an inhibitor that targets Aminoimidazole carboxamide ribonucleotide transformylase/inosine monophosphate cyclohydrolase (ATIC) dimerization with a Ki value of 685 nM. ATIC catalyzes de novo purine biosynthesis, and its dimerization is crucial for AICAR transformylase activity. This inhibitor demonstrates anti-tumor activity by reducing cell numbers and cell division rates and inhibits the proliferation of MCF-7 cells.
  • Targets ATIC dimerization with a Ki value of 685 nM
  • Inhibits proliferation of MCF-7 cells
  • Reduces cell numbers and cell division rates
  • Suitable for studying respiratory diseases, cancer, pulmonary disease, and lung fibrosis
Catalog No. 50-003-75569 Supplier Medchemexpress LLC Supplier No. HY150252A50MG
ATIC-IN-1 acetate is an inhibitor that targets Aminoimidazole carboxamide ribonucleotide transformylase/inosine monophosphate cyclohydrolase (ATIC) dimerization with a Ki value of 685 nM. ATIC catalyzes de novo purine biosynthesis, and its dimerization is crucial for AICAR transformylase activity. This inhibitor demonstrates anti-tumor activity by reducing cell numbers and cell division rates and inhibits the proliferation of MCF-7 cells.
  • Targets ATIC dimerization with a Ki value of 685 nM
  • Inhibits proliferation of MCF-7 cells
  • Reduces cell numbers and cell division rates
  • Suitable for studying respiratory diseases, cancer, pulmonary disease, and lung fibrosis