Learn More
Medchemexpress LLC Aticaprant (CERC-501) | 1174130-61-0 | MFCD22572355 | 99.6% | 418.50 g/mol | C26H27FN2O2 | 25 MG
SDP
Catalog No. 5000245549
Encompass_Preferred
Aticaprant is a potent, centrally penetrant kappa opioid receptor (KOR) antagonist for research use, exhibiting high affinity (Ki 0.807 nM). It is supplied as a white to off-white solid with documented purity and physicochemical data, and is characterized for both in vitro and in vivo studies and formulation protocols.
Catalog No. 50-002-45549 Supplier Medchemexpress LLC Supplier No. HY10171825MG
Aticaprant is a potent, centrally penetrant kappa opioid receptor (KOR) antagonist for research use, exhibiting high affinity (Ki 0.807 nM). It is supplied as a white to off-white solid with documented purity and physicochemical data, and is characterized for both in vitro and in vivo studies and formulation protocols.