Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ aTIP Beckman™ Biomek™ Pipette Tips, 30μL

Catalog No. 12111066 Shop All OHAUS Products
Change view
Click to view available options
Volume (Metric):
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12-111-066 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-111-066 Supplier BIOTIX™ Supplier No. 63300098

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Beckman Biomek FX/NX, AP384, 384-Head platforms.

  • Made from clear polypropylene
  • With graduation marks
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Agilent/Velocity 11 VPrep and Bravo, 384 Tip Format, Beckman Biomek/Multimek with 384 Head
Graduated Yes
Product Line Beckman
Material Polypropylene
Shape Conical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.