Learn More
Medchemexpress LLC Atipamezole (hydrochloride) | 104075-48-1 | MFCD00864502 | >99.0% | 248.75 g/mol | C14H17ClN2 | 10MM 1ML
SDP
Catalog No. 5000236611
Encompass_Preferred
Atipamezole hydrochloride is a research reagent that acts as a selective α2-adrenoceptor antagonist (MPV-1248) used to reverse α2-agonist effects and probe adrenergic signaling. It is available in solution and solid formats; a common ready-to-use solution is 10 mM in DMSO (1 mL). The material is provided at high purity and is supplied with safety and handling information for research use.
Catalog No. 50-002-36611 Supplier Medchemexpress LLC Supplier No. HY1238010MM 1ML
Atipamezole hydrochloride is a research reagent that acts as a selective α2-adrenoceptor antagonist (MPV-1248) used to reverse α2-agonist effects and probe adrenergic signaling. It is available in solution and solid formats; a common ready-to-use solution is 10 mM in DMSO (1 mL). The material is provided at high purity and is supplied with safety and handling information for research use.