Learn More
Apexbio Technology LLC Atipamezole hydrochloride 104075-48-1 10mg
SDP
Catalog No. 502240813
Encompass_Preferred
Atipamezole hydrochloride (CAS 104075-48-1) is a potent antagonist of 2-adrenergic receptors exhibiting a Ki value of 1 6 nM By selectively inhibiting the 2-adrenergic signaling pathway it modulates neurotransmitter release within the central nervous system Atipamezole hydrochloride is frequently utilized in neuroscience research to investigate mechanisms of noradrenergic regulation with established applications in reversal of sedative effects and evaluation of cognitive function
This item is not returnable. View return policy
Catalog No. 50-224-0813 Supplier Apexbio Technology LLC Supplier No. B722610
Atipamezole hydrochloride (CAS 104075-48-1) is a potent antagonist of 2-adrenergic receptors exhibiting a Ki value of 1 6 nM By selectively inhibiting the 2-adrenergic signaling pathway it modulates neurotransmitter release within the central nervous system Atipamezole hydrochloride is frequently utilized in neuroscience research to investigate mechanisms of noradrenergic regulation with established applications in reversal of sedative effects and evaluation of cognitive function