Learn More
Selleck Chemical LLC Atipamezole hydrochloride 10mg 104075-48-1 Antisedan, MPV-1248 hydrochloride, MPV1248 hydrochloride, MPV 1248 hydrochloride
SDP
Catalog No. 501364107
Encompass_Preferred
Atipamezole (Antisedan, MPV1248) is a synthetic ?2 adrenergic receptor antagonist. It has also been researched in humans as a potential anti-Parkinsonian drug. Atipamezole hydrochloride is hydrochloride form of atipamezole. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4107 Supplier Selleck Chemical LLC Supplier No. S464910MG
Atipamezole (Antisedan, MPV1248) is a synthetic ?2 adrenergic receptor antagonist. It has also been researched in humans as a potential anti-Parkinsonian drug. Atipamezole hydrochloride is hydrochloride form of atipamezole. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.