Learn More
Selleck Chemical LLC Atipamezole hydrochloride S4649-50mg
SDP
Catalog No. 503112128
Encompass_Preferred
Atipamezole (Antisedan MPV1248) is a synthetic 2 adrenergic receptor antagonist It has also been researched in humans as a potential anti-Parkinsonian drug Atipamezole hydrochloride is hydrochloride form of atipamezole
Catalog No. 50-311-2128 Supplier Selleck Chemical LLC Supplier No. S464950MG
Atipamezole (Antisedan MPV1248) is a synthetic 2 adrenergic receptor antagonist It has also been researched in humans as a potential anti-Parkinsonian drug Atipamezole hydrochloride is hydrochloride form of atipamezole