Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atiprimod dihydrochloride, Tocris Bioscience™
SDP

Catalog No. 458010
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-801-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-801-0 Supplier R&D Systems Supplier No. 4580/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

JAK2 inhibitor

JAK2 inhibitor (IC50 = 397 nM). Also inhibits STAT3 and STAT5 phosphorylation. Inhibits cell growth and induces apoptosis in cells expressing the JAK2V617F mutation.

Chemical Identifiers

CAS 130065-61-1
Molecular Formula C22H46Cl2N2
Molecular Weight (g/mol) 409.524
InChI Key MOUZYBYTICOTFQ-UHFFFAOYSA-N
Synonym atiprimod dihydrochloride, atiprimod dihydrochloride usan, atiprimod hcl, atiprimod hydrochloride, 2-3-diethylamino propyl-8,8-dipropyl-2-azaspiro 4.5 decane dihydrochloride, 2-azaspiro 4.5 decane-2-propanamine n,n-diethyl-8,8-dipropyl-, dihydrochloride, 2-azaspiro 4.5 decane-2-propanamine, n,n-diethyl-8,8-dipropyl-, dihydrochloride, 3-8,8-dipropyl-3-azaspiro 4.5 decan-3-yl-n,n-diethylpropan-1-amine dihydrochloride, n,n-diethyl-8,8-dipropyl-2-azaspiro 4.5 decane-2-propanamine dihydrochloride
PubChem CID 166556
IUPAC Name 3-(8,8-dipropyl-2-azaspiro[4.5]decan-2-yl)-N,N-diethylpropan-1-amine;dihydrochloride
SMILES CCCC1(CCC2(CC1)CCN(C2)CCCN(CC)CC)CCC.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 409.52
Chemical Name or Material Atiprimod dihydrochloride
Recommended Storage Desiccate at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.