Learn More
Cooper Atkins Corporation ATKINS PROBE 50209K 1/EA
Catalog No. NC9199604
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
MicroNeedle, Technology Base: Thermocouple, Thermocouple Type: Type K, Max Tip Temperature: 500degreeF/260degreeC, Max Cord Temperature: 176degreeF/80degreeC, Response Time: 1 sec. response in liquids, Shaft Length: 3.5 in./89mm
Catalog No. NC9199604 Supplier Cooper Atkins Corporation Supplier No. 50209K
This item has a minimum qty of 2 per supplier requirements.
MicroNeedle, Technology Base: Thermocouple, Thermocouple Type: Type K, Max Tip Temperature: 500degreeF/260degreeC, Max Cord Temperature: 176degreeF/80degreeC, Response Time: 1 sec. response in liquids, Shaft Length: 3.5 in./89mm