Learn More
Biotage ATL DISK DVB 50MM-BX24 EA=20
SDP
Catalog No. NC0200991
Encompass
Atlantic DVB (divinylbenzene) polymer is a pH independent reversed-phase media that is listed for use in EPA Method 525.3. The Atlantic DVB Disk has also proven effective for the analysis of EPA Method 8270 and 1694, endocrine disruptors, personal care products, pharmaceutical residues, and other emerging contaminants in surface waters and drinking waters. Resistant to accidental overdrying.
Catalog No. NC0200991 Supplier Biotage Supplier No. 47234606
Atlantic DVB (divinylbenzene) polymer is a pH independent reversed-phase media that is listed for use in EPA Method 525.3. The Atlantic DVB Disk has also proven effective for the analysis of EPA Method 8270 and 1694, endocrine disruptors, personal care products, pharmaceutical residues, and other emerging contaminants in surface waters and drinking waters. Resistant to accidental overdrying.