Learn More
Aviva Systems Biology Corp Atl2 Antibody - C-terminal region (ARP49719_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49719_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Atl2. It was validated on Western Blot. . Alternate names: 2010110I21Rik, AA407293, AV334690, Aip-2, Arl6ip2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-446