Learn More
ABclonal Technology ATL2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A158841000UL
Encompass_Preferred
Enables identical protein binding activity. Involved in Golgi organization, endoplasmic reticulum tubular network membrane organization, and protein homooligomerization. Located in endoplasmic reticulum tubular network membrane. Is integral component of membrane.
Catalog No. 50-001-06569