Learn More
Biotage ATLANTIC DISK C18 HC 50MM 24PK
SDP
Catalog No. NC0469682
Encompass
Atlantic high capacity c18 spe disk provides added retention per disk for improved recovery of analytes from higher organic content sampes and improved recovery of compounds that have weaker interactions with c18 media. much like the original atlantic c18, the new high capactiy c18 can be used for drinking water applications such as epa method 525.2.
Catalog No. NC0469682 Supplier Biotage Supplier No. 47234613
Atlantic high capacity c18 spe disk provides added retention per disk for improved recovery of analytes from higher organic content sampes and improved recovery of compounds that have weaker interactions with c18 media. much like the original atlantic c18, the new high capactiy c18 can be used for drinking water applications such as epa method 525.2.