Learn More
Biotage ATLANTIC HLB DISK-M 30PK
SDP
Catalog No. NC0167026
Encompass
Atlantic HLB Medium Capacity SPE Disk is designed for samples with medium organic content and extracting EPA 8270,8081,8082 (semi-volatile organic compounds) and EPA 1694 ( Pharmaceutical By-Products). This is a great in.all purpose in. disk which provides excellent extraction of a large range of semi-volatile compounds.
Catalog No. NC0167026 Supplier Biotage Supplier No. 47234609
Atlantic HLB Medium Capacity SPE Disk is designed for samples with medium organic content and extracting EPA 8270,8081,8082 (semi-volatile organic compounds) and EPA 1694 ( Pharmaceutical By-Products). This is a great in.all purpose in. disk which provides excellent extraction of a large range of semi-volatile compounds.