Learn More
Biotage Atlantic® ReadyDisk C18, Single-use Solid Phase Extraction (SPE) Disks for Water Extraction, 24 per Pack
SDP
Catalog No. NC2825304
Encompass
Atlantic® ReadyDisks are solid phase extraction (SPE) disks, pre-assembled into disposable holders and used for extracting semivolatile organic compounds from aqueous matrices. The ReadyDisks are designed for single-use extractions with maximum speed and convenience in mind.Each disk is constructed to maximize surface area interactions with the analytes in solution, accommodating relatively high sample flow rates without suffering from solution or analyte breakthrough or disk clogging.The disk holder was designed for use with both manual and automated solid phase extraction systems and is constructed from materials that are free from contaminants such as phthalates. The configuration of the disk results in high physical and chemical capacity and the height of the holder prevents sample overflow, even at relatively high flow rates.
Catalog No. NC2825304 Supplier Biotage Supplier No. 476005
Atlantic® ReadyDisks are solid phase extraction (SPE) disks, pre-assembled into disposable holders and used for extracting semivolatile organic compounds from aqueous matrices. The ReadyDisks are designed for single-use extractions with maximum speed and convenience in mind.Each disk is constructed to maximize surface area interactions with the analytes in solution, accommodating relatively high sample flow rates without suffering from solution or analyte breakthrough or disk clogging.The disk holder was designed for use with both manual and automated solid phase extraction systems and is constructed from materials that are free from contaminants such as phthalates. The configuration of the disk results in high physical and chemical capacity and the height of the holder prevents sample overflow, even at relatively high flow rates.