Learn More
Kingfisher Biotech Inc Atlantic Salmon IGF-1 Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Catalog No. 502964769
Encompass
The Atlantic Salmon IGF-1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Atlantic Salmon IGF-1 applications are for cell culture, ELISA standard, and Western Blot Control. Atlantic Salmon IGF-1 Specifications: (Molecular Weight: 15.1 kDa) (Amino Acid Sequence: ATGAGPETLC GAELVDTLQF VCGERGFYFS KPTGYGPSSR RSHNRGIVDE CCFQSCELRR LEMYCAPVKS GKAARSVRAQ RHTDMPRTPK VSTAVQNVDR GTERRTAQHP DKTKTKKEVH QKNSSRGNTG GRNYRM). For research use only. Made in the USA
Catalog No. 50-296-4769 Supplier Kingfisher Biotech Inc Supplier No. RP2171AS025
The Atlantic Salmon IGF-1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Atlantic Salmon IGF-1 applications are for cell culture, ELISA standard, and Western Blot Control. Atlantic Salmon IGF-1 Specifications: (Molecular Weight: 15.1 kDa) (Amino Acid Sequence: ATGAGPETLC GAELVDTLQF VCGERGFYFS KPTGYGPSSR RSHNRGIVDE CCFQSCELRR LEMYCAPVKS GKAARSVRAQ RHTDMPRTPK VSTAVQNVDR GTERRTAQHP DKTKTKKEVH QKNSSRGNTG GRNYRM). For research use only. Made in the USA