Learn More
Waters Corp Atlantis Method Validation Kit, BEH C18 AX, 2.5 μm 2.1 x 50 mm
Catalog No. NC1825043 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns, each packed with a different lot of material, designed for method development. It provides excellent retention for polar acidic analytes under reversed-phase conditions. The reversed-phase/anion-exchange mixed-mode chemistry is stable from pH 2 to 10 and offers complementary selectivity to traditional C18 stationary phases.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Features reversed-phase/anion-exchange mixed-mode chemistry, stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases, ideal for method development.
  • Utilizes BEH particle technology with a 2.5 μm particle size and 95 Å pore size.
  • Compatible with UPLC and UHPLC systems, supporting liquid chromatography (LC) applications.
Catalog No. NC1825043 Supplier Waters Corp Supplier No. 186009394
This method validation kit includes three columns, each packed with a different lot of material, designed for method development. It provides excellent retention for polar acidic analytes under reversed-phase conditions. The reversed-phase/anion-exchange mixed-mode chemistry is stable from pH 2 to 10 and offers complementary selectivity to traditional C18 stationary phases.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Features reversed-phase/anion-exchange mixed-mode chemistry, stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases, ideal for method development.
  • Utilizes BEH particle technology with a 2.5 μm particle size and 95 Å pore size.
  • Compatible with UPLC and UHPLC systems, supporting liquid chromatography (LC) applications.